Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cis-benzyl 3-hydroxycyclobutylcarbamate | 1403766-86-8 | MFCD23106304 | 221.26 g/mol | C12H15NO3 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW05781110G

Encompass_Preferred

Cis-benzyl 3-hydroxycyclobutylcarbamate is an organic drug intermediate used in medicinal chemistry for the synthesis of active compounds. It is a solid with molecular formula C12H15NO3 and molecular weight 221.26 g/mol; CAS number 1403766-86-8. Recommended storage is 4°C, sealed and protected from moisture; in solvent: -80°C for up to 6 months, or -20°C for up to 1 month.

  • Used as a drug intermediate in medicinal chemistry syntheses.
  • Solid form for standard laboratory handling.
  • Molecular formula C12H15NO3 and molecular weight 221.26 g/mol.
  • Recommended storage: 4°C, sealed and protected from moisture; in solvent: -80°C (6 months) or -20°C (1 month).
  • Available as a 10 G package.

Catalog No. 50-002-10789


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.