Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cis-benzyl 3-hydroxycyclobutylcarbamate | 1403766-86-8 | MFCD23106304 | 221.26 g/mol | C12H15NO3 | 50 G
SDP

Supplier:  Medchemexpress LLC HYW05781150G

Encompass_Preferred

cis-Benzyl 3-hydroxycyclobutylcarbamate is a small-molecule drug intermediate used in medicinal chemistry for the synthesis of active pharmaceutical compounds. It has molecular formula C12H15NO3 and molecular weight 221.26 g/mol. Handle according to the safety data sheet: harmful if swallowed, store sealed and protected from moisture at 4°C.

  • Used as a synthetic intermediate in medicinal chemistry
  • Molecular formula C12H15NO3
  • Molecular weight 221.26 g/mol
  • Recommended storage: 4°C, sealed, away from moisture
  • Hazard: harmful if swallowed (GHS acute toxicity category 4); follow SDS precautions

Catalog No. 50-002-10036


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.