Learn More
Medchemexpress LLC Cis-benzyl 3-hydroxycyclobutylcarbamate | 1403766-86-8 | MFCD23106304 | 97.0% | 221.26 | C12H15NO3 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW057811
cis-Benzyl 3-hydroxycyclobutylcarbamate is a small-molecule drug intermediate used in the synthesis of active pharmaceutical compounds. It is supplied as a solid and offered in multiple pack sizes for research and development applications.
- Used as a chemical intermediate for synthesis of active compounds.
- Molecular formula C12H15NO3; molecular weight 221.26.
- CAS number 1403766-86-8 for unambiguous identification.
- Available in multiple pack sizes, including 250 mg, 500 mg, 1 g, and larger quantities.
- Recommended storage: 4°C, sealed and away from moisture; in solvent: -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.