Learn More
Strem, An Ascensus Company CAS# 113822-11-0. 1g. cis-Dimethyl(N,N,N',N'-tetramethylethylenediamine)palladium(II), 99%. MFCD11045096
SDP
Supplier:  Strem, An Ascensus Company 4609521G
Encompass
CAS# 113822-11-0. 1g. cis-Dimethyl(N,N,N',N'-tetramethylethylenediamine)palladium(II), 99%. MFCD11045096. Molecular Weight: 252.69. Molecular Formula: C8H22N2Pd. Color/form: white to off-white pwdr. Strem# 46-0952. http://www.strem.com/catalog/v/46-0952/
Catalog No. 50-901-12366