Learn More
Apexbio Technology LLC cis-Epoxysuccinic acid 16533-72-5 1g
SDP
Supplier:  Apexbio Technology LLC A99701000
Encompass_Preferred
cis-Epoxysuccinic acid (CAS 16533-72-5) is an epoxysuccinic acid derivative that functions as an agonist of the succinate receptor SUCNR1 (GPR91) Activation of SUCNR1 by cis-Epoxysuccinic acid leads to a marked decrease in intracellular cyclic AMP (cAMP) concentrations with an EC50 of approximately 2 7 M This modulation of cAMP signaling may impact various cellular processes including metabolic regulation and inflammatory responses cis-Epoxysuccinic acid serves as a useful tool compound for investigating SUCNR1-mediated pathways and cAMP-associated signaling in cellular and molecular research
Catalog No. 50-000-95910