Learn More
Medchemexpress LLC Cis-Fmoc-Pro(4-N3)-OH | 263847-08-1 | MFCD04115792 | 99.1% | 378.38 g/mol | C20H18N4O4 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW142062250MG
Encompass_Preferred
cis-Fmoc-Pro(4-N3)-OH is an Fmoc-protected proline derivative carrying an azide at the 4-position. Supplied as a white to off-white solid, it is intended as a building block for peptide synthesis and for introducing azide functionality for bioorthogonal conjugation via copper-catalyzed azide-alkyne cycloaddition.
  • Contains azide functional group for copper-catalyzed azide-alkyne cycloaddition (CuAAC) conjugation.
  • Fmoc-protected for compatibility with standard solid-phase peptide synthesis workflows.
  • High purity suitable for synthetic and analytical applications.
  • Stable as a powder when stored under recommended conditions for long-term use.
  • Available in multiple sizes, including small 250 mg quantities for research use.
Catalog No. 50-002-83786