Learn More
Medchemexpress LLC Cis-[Pt(NH3)2(phenanthridine)Cl]NO3 | 1416900-51-0 | 95.0% | C13H13ClN4O3Pt | 10MG
SDP
Supplier:  Medchemexpress LLC HY13981310MG
Encompass_Preferred
Phenanthriplatin (cis-[Pt(NH3)2(phenanthridine)Cl]NO3) is a monovalent platinum(II) complex with potent cytotoxic activity against cancer cells. Supplied as a solid for laboratory research, it is intended for in vitro and mechanistic studies of DNA-targeting platinum therapeutics. For research use only; not for human or clinical use.
  • Monovalent platinum(II) complex with DNA-binding properties.
  • Suitable for in vitro cytotoxicity and mechanism-of-action studies.
  • Supplied as a white to off-white solid for convenient handling.
  • Typical purity reported by supplier: 95.0%.
  • Stable when stored sealed, away from moisture and light; solvent storage guidance available.
  • Available in small research-scale packaging for experimental workflows.
Catalog No. 50-002-60432