Learn More
Abcam cis-Resveratrol, Antio x idant agent, 100MG
Supplier:  Abcam AB144436100MG
Encompass_Preferred
MW 228.24 Da, Purity >98%. Antioxidant agent. NADPH oxidase inhibitor. Blocks ROS production. Resveratrol (ab120726) isomer. Inhibits canonical and non-canonical inflammasomes in macrophages. Shows anti-inflammatory and antiproliferative effects. Orally bioavailable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5730