Learn More
Bioworld cis,trans-Abscisic Acid (ABA), 250 mg
Supplier:  Bioworld 306310171
Encompass_Preferred
(+)- cis, trans-Abscisic acid (ABA)Specifications:A plant hormone involved in plant growth and stress responsePlant cell culture testedAppearance: white to off-white powderMelting Point: 186-188°CSoluble in methanolDo not expose to a direct light source
Catalog No. 50-255-068