Learn More
Medchemexpress LLC Cis-urocanic acid | 7699-35-6 | 99.9% | 138.12 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY113008A1MG
Encompass_Preferred
cis-Urocanic acid is a 5-HT2A receptor agonist that binds to the 5-HT receptor with relatively high affinity (Kd=4.6 nM). This compound acts as an immune modulator, inducing immunosuppression through its binding to the 5-HT2A receptor.
  • 5-HT2A receptor agonist.
  • Immune modulator that induces immunosuppression by binding to the 5-HT2A receptor.
  • Treatment with 100 μg/mL completely suppresses IL-6 and IL-8 secretion.
  • Decreases caspase-3 activity.
  • Improves cell viability against UV-B irradiation in human corneal epithelial cells (HCE-2) and human conjunctival epithelial cells (HCECs).
Catalog No. 50-004-39853