Learn More
Medchemexpress LLC Cis-Urocanic acid | 7699-35-6 | 99.9% | 138.12 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY113008100MG
Encompass_Preferred
cis-Urocanic acid is a 5-HT2A receptor agonist that binds to the 5-HT receptor with a relatively high affinity (Kd=4.6 nM). It acts as an immune modulator, inducing immunosuppression by binding to the 5-HT2A receptor. This product is for research use only and not sold to patients.
  • Binds to the 5-HT2A receptor with high affinity
  • Induces immunosuppression
  • Suppresses IL-6 and IL-8 secretion
  • Decreases caspase-3 activity
  • Improves cell viability against UV-B irradiation
Catalog No. 50-004-39403