Learn More
Medchemexpress LLC Cis VH-298 | 2097381-86-5 | 100.0% | 523.65 g/mol | C27H33N5O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY100947A10MG
Encompass_Preferred
cis VH-298 is the cis stereoisomer of VH-298 that acts as a VHL:HIF-α interaction inhibitor. Compared with the trans isomer, the cis form shows a significant loss of binding specificity of hydroxylated HIF-1α peptides to the VHL protein in vitro. The compound is supplied as a white to off-white solid with high purity for research use and should be stored protected from light.
  • Cis stereochemistry reduces binding specificity to VHL.
  • Useful as a comparator in VHL-HIF biological studies.
  • High purity (approximately 99.96%) suitable for biochemical assays.
  • Molecular weight 523.65 g/mol; formula C27H33N5O4S.
  • Solid, white to off-white appearance; store protected from light.
Catalog No. 50-002-12622