Learn More
Medchemexpress LLC Cisapride | 81098-60-4 | 99.9% | C23H29ClFNO4 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY14149200MG
Encompass_Preferred
Cisapride is an orally active 5-HT4 receptor agonist with an EC50 value of 140 nM. It is also a hERG blocker with an IC50 value of 9.4 nM. This gastroprokinetic agent stimulates gastrointestinal motor activity and is intended for research use only.
  • Orally active 5-HT4 receptor agonist
  • hERG blocker
  • Stimulates gastrointestinal motor activity
  • Suitable for high-speed applications
  • Available in various solid pack sizes
Catalog No. 50-003-56809