Learn More
Medchemexpress LLC HY-W000800 500mg Medchemexpress, cis,cis-Muconic acid CAS:1119-72-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW000800500MG
Encompass_Preferred
Medchemexpress, HY-W000800 500mg cis,cis-Muconic acid CAS:1119-72-8 cis,cis-Muconic acid, a metabolic intermediate of Klebsiella pneumonia, can be converted to adipic acid and terephthalic acid, which are important monomers of synthetic polymers. cis,cis-Muconic acid is also a biochemical material that can be used for the production of various plastics and polymers and is particularly gaining attention as an adipic acid precursor for the synthesis of nylon-6,6. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3199