Learn More
Medchemexpress LLC Cisplatin | 15663-27-1 | 99.3% | 10 MG

Supplier: Medchemexpress LLC HY1739410MG
Cisplatin is a platinum-based antineoplastic research reagent that cross-links DNA and causes DNA damage, leading to cell-cycle arrest, apoptosis, ferroptosis, and autophagy. It is provided as a laboratory-grade solid with documented solubility profiles and storage recommendations for in vitro and in vivo applications.
- Acts as a DNA alkylator and crosslinker.
- Induces apoptosis, ferroptosis, and autophagy in cancer cells.
- High reported purity (≈99.3%).
- Soluble in DMF and partially soluble in water with warming and ultrasonic treatment.
- Suitable for both in vitro and in vivo formulations with appropriate vehicle preparation.
- Store protected from light at 4°C; in solution store at -20°C to -80°C for extended periods.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.