Learn More
AdipoGen Cisplatin, 15663-27-1, MFCD00011623, 50 mg
SDP
Supplier:  AdipoGen AGCR13590M050
Encompass_Preferred
MF: Cl2H6N2Pt, MW: 300.0, ≥98%, Appearance: Yellow solid. Potent platinum-based antineoplastic agent, Anticancer compound, Forms inter- and intrastrand DNA adducts/crosslinks, consequently blocking DNA replication and transcription and inducing cell death, Nephrotoxic when used in vivo, and potent chemoimmunotherapeutic. Stimulates the immune responses by activating macrophages and other cells of the immune system.
Catalog No. 50-595-860