Learn More
Selleck Chemical LLC Cisplatin S1166-10g
SDP
Supplier:  Selleck Chemical LLC S116610G
Encompass_Preferred
Cisplatin is an inorganic platinum complex which is able to inhibit DNA synthesis by conforming DNA adducts in tumor cells Cisplatin activates ferroptosis and induces autophagy Solutions are unstable and should be fresh-prepared DMSO is not recommended to dissolve platinum-based drugs which can easily lead to drug inactivation
Catalog No. 50-311-1176