Learn More
Selleck Chemical LLC Cisplatin 50mg 15663-27-1 cisplatinum, cis-diamminedichloroplatinum II, CDDP
SDP
Supplier:  Selleck Chemical LLC S116650MG
Encompass_Preferred
Cisplatin (cisplatinum, cis-diamminedichloroplatinum II, CDDP) is an inorganic platinum complex, which is able to inhibit DNA synthesis by conforming DNA adducts in tumor cells. Cisplatin activates ferroptosis and induces autophagy. Solutions are best fresh-prepared. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0564900