Learn More
Medchemexpress LLC Citalopram | 59729-33-8 | 99.8% | 100 MG

Supplier: Medchemexpress LLC HY121203100MG
Citalopram (CAS 59729-33-8) is a racemic selective serotonin reuptake inhibitor (SSRI) provided as an analytical research standard for laboratory use. It is intended for analytical and pharmacological research and method development; not for human or clinical use.
- Analytical standard for research use only.
- Molecular formula C20H21FN2O.
- Molecular weight 324.39 g/mol.
- Reported purity about 99.8% (manufacturer listing; may vary by batch).
- Supplied as a 100 MG quantity suitable for analytical assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.