Learn More
TARGETMOL CHEMICALS INC Citalopram hydrobromide 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1483100MG
Encompass_Preferred
Also available in 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Citalopram hydrobromide (XU-62-320) , a selective serotonin reuptake inhibitor (SSRI), selectively inhibits the CNS neuronal reuptake of serotonin, thereby potentiating serotonergic activity in the central nervous system (CNS). Citalopram hydrobromide is the orally bioavailable hydrobromide salt of the racemic bicyclic phthalene derivative citalopram with antidepressant activity. This agent has minimal effects on the CNS neuronal reuptake of norepinephrine (NE) and dopamine (DA). Purity 99.35%
Catalog No. 50-287-8535