Learn More
Apexbio Technology LLC Citalopram hydrobromide 59729-32-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B67365.1
Encompass_Preferred
Citalopram hydrobromide (CAS 59729-32-7) is a selective serotonin reuptake inhibitor (SSRI) that functions by specifically blocking the reuptake of 5-hydroxytryptamine (5-HT) at presynaptic neuronal membranes thereby increasing 5-HT concentrations in the synaptic cleft In vitro studies demonstrate that citalopram hydrobromide exhibits potent nanomolar-level inhibition of 5-HT reuptake in synaptosomal preparations as well as low nanomolar inhibition in rabbit platelet assays This compound is widely utilized in research exploring serotonergic signaling neurotransmission processes and experimental models of depression
Catalog No. 50-000-91785