Learn More
Medchemexpress LLC Citco | 338404-52-7 | MFCD00139608 | 99.6% | 436.74 | C19H12Cl3N3OS | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10324410MG
Encompass_Preferred
CITCO is an imidazothiazole-derived small molecule that functions as a selective agonist of the constitutive androstane receptor (CAR), used in research to probe CAR-mediated signaling pathways and pharmacology. It is supplied as a solid reagent with high purity for in vitro studies.
  • Selective CAR agonist for receptor activation studies.
  • High purity suitable for research applications.
  • Solid, white to light yellow appearance for easy handling.
  • Storage: solid at -20°C; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small research sizes, including 10 mg.
Catalog No. 50-002-46189