Learn More
Medchemexpress LLC Citraconimide | 1072-87-3 | MFCD10699598 | 96.8% | 111.10 g·mol⁻1 | C5H5NO2 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW023557
Encompass_Preferred
Citraconimide (CAS 1072-87-3) is a 3-methylmaleimide derivative used as a thiol-reactive reagent and building block in organic synthesis. It acts as a Michael acceptor for regioselective thiol additions and is supplied with analytical characterization suitable for research use.
  • Thiol-selective Michael acceptor for conjugation chemistry.
  • Effective building block for small-molecule synthesis and modification.
  • Regioselective addition with a variety of alkyl and aryl thiols.
  • Compatible with common organic solvents and mild reaction conditions.
  • Provided with lot-specific analytical data and purity specification.
Catalog No. 50-003-03748