Learn More
Biogenex Laboratories CITRATE BUFFER PH 6 250ML
SDP
Supplier:  Biogenex Laboratories HK0875K
Encompass_Preferred
Antigen Retrieval Citra Solution 250mL Contains the buffer used with our proprietary microwave Antigen Retrieval method Provides staining of routine paraffin sections with most BioGenex antibodies
Catalog No. NC9571809
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.