Learn More
Bioworld Citrate-MOPS Buffer, 10 PK(s)
Supplier:  Bioworld 402200801
Encompass_Preferred
Product DescriptionCitrate-MOPS (3-(N-morpholino)propanesulfonic acid) buffer comes in a quantity of 10 packs, each pack yields 0.6M sodium citrate and 0.1M MOPS buffer, at a pH of 7.5. Dissolve in 100ml of deionized water to make 1 liter.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.
Catalog No. 50-255-477