Learn More
Medchemexpress LLC Citrazinic acid | 99-11-6 | MFCD00006274 | ≥90.0% | 155.11 g/mol | C6H5NO4 | 10 G

Supplier: Medchemexpress LLC HYW02581710G
Citrazinic acid (2,6-dihydroxyisonicotinic acid) is a fluorescent pyridine carboxylic acid used as a research reagent in analytical and spectroscopic applications. Supplied as a solid, it exhibits concentration- and environment-dependent optical properties useful for fluorescence studies.
- Fluorescent pyridine carboxylic acid.
- Molecular formula C6H5NO4; molecular weight 155.11 g/mol.
- Purity ≥90.0%.
- Available in small laboratory pack sizes (10 g, 25 g).
- Suitable for spectroscopy and fluorescence studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.