Learn More
Selleck Chemical LLC Citric acid monohydrate E2164-250MG
SDP
Supplier:  Selleck Chemical LLC E2164250MG
Encompass_Preferred
Citric acid monohydrate is a natural preservative and food tartness enhancer Citric acid monohydrate induces apoptosis and cell cycle arrest at G2/M phase and S phase Citric acid monohydrate cause oxidative damage of the liver by means of the decrease of antioxidative enzyme activities Citric acid monohydrate causes renal toxicity in mice
Catalog No. 50-000-39202