Learn More
Enzo Life Sciences Citrinin (5mg). CAS: 518-75-2
SDP
Supplier:  Enzo Life Sciences BMLCM1160005
Encompass_Preferred
Antibiotic. Induces mitochondrial permeability pore opening and inhibits respiration by interfering with complex I of the respiratory chain. Mycotoxin. Acts as nephrotoxin in all species in which it has been tested. Has been implicated as a cause of Balkan nephropathy and yellow rice fever in humans. Induces apoptosis. Purity: ≥98% (HPLC, TLC). Solubility: Soluble in 100% ethanol, methylene chloride (10mg/ml), methanol, dioxane, pyridine or dichlormethane. Insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-0420