Learn More
Cayman Chemical CItrulInATd HIston H3hum 100ug
SDP
Supplier:  Cayman Chemical 17926100 UG
Encompass_Preferred
Source: Recombinant human histone H3 expressed in E. coli and citrullinated by PAD4 • Amino acids: 1-136 • MW: 15.5 kDa
Catalog No. 50-296-5976