Learn More
Cayman Chemical CItrulInATd Hsp70humn re 100ug
SDP
Supplier:  Cayman Chemical 25108100 UG
Encompass_Preferred
Source: N-Terminal histidine-tagged human Hsp70 purified from E. coli, citrullinated by PAD2 • MW: 71.7 kDa
Catalog No. 50-293-2515