Learn More
Bachem H-Cit-AMC · HBr
SDP
Supplier:  Bachem 4019017.0100
Encompass_Preferred
H-Cit-AMC · HBr, 123314-39-6, C16H20N4O4 · HBr, Mr 413.27, 100mg. H-Orn(carbamoyl)-AMC · HBr. Citrulline-MCA is most efficiently hydrolyzed by both human and yeast bleomycin hydrolase.
Catalog No. 50-194-7007