Learn More
Cayman Chemical CItrus maxa peel MernzIn 5mg
SDP
Supplier:  Cayman Chemical 347435 MG
Encompass_Preferred
A coumarin with diverse biological activities; cytotoxic to NSCLC-N6 and KB cancer cells (IC50s = 14.6 and 12.3 UG/ml, respectively); increases the latency to paw withdrawal in the hot plate test and reduces xylene-induced EAr edema in mice at 10 mg/kg; decreases carrageenan-induced paw edema in rats at 10 mg/kg
Catalog No. 50-291-2449