Learn More
Medchemexpress LLC Cjoc42 | 2171519-89-2 | 99.0% | 415.46 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13805450MG
Encompass_Preferred
Cjoc42 is a compound capable of binding to gankyrin, inhibiting its activity in a dose-dependent manner. It prevents the decrease in p53 protein levels normally associated with high amounts of gankyrin, thereby restoring p53-dependent transcription and sensitivity to DNA damage.
  • Capable of binding to gankyrin
  • Inhibits gankyrin activity dose-dependently
  • Restores p53-dependent transcription
  • Restores sensitivity to DNA damage
Catalog No. 50-003-51684