Learn More
Selleck Chemical LLC CK-586 E4662-100MG
SDP
Supplier:  Selleck Chemical LLC E4662100MG
Encompass_Preferred
CK-586(CK-4021586) is an inhibitor of cardiac myosin used to treat obstructive hypertrophic cardiomyopathy (oHCM) It effectively reduces left ventricular outflow tract obstruction (LVOTO) in a dose-dependent manner
Catalog No. 50-000-39438