Learn More
Selleck Chemical LLC CK-666
SDP
Supplier:  Selleck Chemical LLC S769025MG
Encompass_Preferred
CK-666 is an inhibitor of the actin-related protein 2/3 (Arp2/3) complex with IC50 of 17 M and 5 M for BtArp2/3 complex and SpArp2/3 complex respectively
Catalog No. 50-000-00432