Learn More
Selleck Chemical LLC CK-869
SDP
Supplier:  Selleck Chemical LLC S69545MG
Encompass_Preferred
CK-869 (CK 0157869) is a small molecule inhibitor of actin-related protein 2/3 (Arp2/3) complex with an IC50 of 7 M CK-869 can directly suppresses microtubule assembly both in vitro and in vivo
Catalog No. 50-000-02244