Learn More
Sino Biological Peptide Substrates: CK1tide
Supplier:  Sino Biological C07581
Encompass_Preferred
The CK1tide synthetic peptide [HAAIGDDDDAYSITA-NH2] is routinely evaluated as a substrate for CK1 family kinases, such as CK1 and CK1 delta.
Catalog No. 50-256-9630