Learn More
Cayman Chemical cKItCD117CTerm RabbIt M 100uL
SDP
Supplier:  Cayman Chemical 32286100 UL
Encompass_Preferred
An internal standard for the quantification of (4-carboxybutyl)triphenylphosphonium by GC- or LC-MS
Catalog No. 50-295-0786