Learn More
Foxx Life Sciences Autofil Sterile Polystyrene (PS) Media Storage Bottles with SECUREgrasp GL45 Cap, 500mL, 24/CS
Supplier:  Foxx Life Sciences 1172RLS
Encompass_Preferred
1172-RLS: Media Bottle, 500 mL, PS, Sterile. Foxx Life Sciences' Polystyrene Media Bottles have been designed specifically for Life Science researchers to store common solutions such as tissue culture media, serum, and buffers. Our bottles are designed to handle full vacuum, and can be used with the Autofil filtration system. Our Polysytrene bottles are used for general purpose cell culture aqueous solutions. Each Autofil bottle has features that improve the handling of the bottle, especially when the bottle is wet. The narrow bottle shoulder, combined with the molded finger grips, improves the ability to securely grip the bottle with gloved hands. Our patented, SECUREgrasp bottle cap enables easy handling and carrying of the bottle. The innovative cap also allows for easy uncapping, tightening, and bottle stacking and provides a large surface for bottle labeling. Foxx Life Sciences' Autofil is manufactured in ISO 13485 clean room in North America.
Catalog No. 50-105-0038