Learn More
Cayman Chemical CL 316 243sodIum salt 10mg
SDP
Supplier:  Cayman Chemical 1749910 MG
Encompass_Preferred
A β3-adrenoceptor agonist (EC50 = 3 nM) that is >10,000-fold selective over β1 and β2; causes a rapid decrease in blood glucose level in mice and has been used to study the induction of brown adipocytes and to increase thermogenesis in a mouse model of dietary obesity
Catalog No. 50-296-6882