Learn More
Medchemexpress LLC HY-116771A 10mg , CL 316243 CAS:138908-40-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY116771A10MG
Encompass_Preferred
HY-116771A 10mg CL 316243 CAS: 138908-40-4 CL316243 is a highly potent selective β3-adrenoceptor agonist with a EC50 of 3 nM, but is an extremely poor to β1/2- receptors.CL316243 is a effective stimulant of adipocyte lipolysis and increases brown adipose tissue thermogenesis and metabolic rate. CL316243 has the potential for the treatment obesity, diabetes and urge urinary incontinence. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0576