Learn More
Medchemexpress LLC HY-10325 5mg Medchemexpress, CL-387785 CAS:194423-06-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY103255MG
Encompass_Preferred
Medchemexpress, HY-10325 5mg CL-387785 CAS:194423-06-8 CL-387785(EKI785; WAY-EKI 785) is an irreversible inhibitor of EGFR with IC50 of 370 pM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8061