Learn More
Medchemexpress LLC Cl-C6-PEG4-O-CH2COOH (PROTAC Linker 4) | 1799506-30-1 | 98.0% | 370.87 | 50 MG

Supplier: Medchemexpress LLC HY11249650MG
Cl-C6-PEG4-O-CH2COOH (PROTAC Linker 4) is a PEG-based PROTAC linker used in the synthesis of chloroalkane-containing PROTACs (HaloPROTACs). This product is for research use only. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting an E3 ubiquitin ligase ligand and a target protein ligand via a linker.
- Appearance: oil
- Color: colorless to light yellow
- Chemical formula: C16H31ClO7
- Shipping: room temperature in continental US
- Storage in pure form: -20°C for 3 years, 4°C for 2 years
- Storage in solvent: -80°C for 6 months, -20°C for 1 month
- Solubility in ethanol: 100 mg/mL (needs ultrasonic)
- Solubility in DMSO: 100 mg/mL (needs ultrasonic, use newly opened DMSO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.