Learn More
MSC CLR Pro Max Industrial Degreaser, Water-Based, Concentrated, 5 gal.
Supplier:  MSC 82597105
Encompass
This industrial-grade, water-based degreaser is a concentrated, non-flammable formula designed to effectively penetrate and dissolve heavy grease, oil, and grime from machinery, parts, and heavy equipment. Containing no abrasives, acids, or petroleum solvents, this versatile cleaner is safe for use in various industrial settings, including food processing and service facilities.
  • Concentrated formula allows for adjustable strength based on cleaning requirements
  • NSF registered for use in food processing, food storage, and food service facilities
  • Non-flammable and contains no VOCs
  • Effective on diverse surfaces including concrete, metal, stainless steel, and fiberglass
  • Suitable for pressure washers, dip tanks, and floor scrubbers
  • EPA Safer Choice certified
Catalog No. 50-005-07814