Learn More
TARGETMOL CHEMICALS INC CL097 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T384235MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CL097 is an effective agonist of TLR7 and TLR8. CL097 induces pro-inflammatory cytokines in macrophages and NADPH oxidase priming thereby increasing the fMLF-stimulated ROS production. purity: 99%
Catalog No. 50-258-0106