Learn More
Medchemexpress LLC Cl097 (hydrochloride) | 2990770-48-2 | 100.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY128799A10MM/1ML
Encompass_Preferred
CL097 hydrochloride is a potent agonist for Toll-like Receptor 7 (TLR7) and Toll-like Receptor 8 (TLR8). It induces the production of pro-inflammatory cytokines in macrophages and also triggers NADPH oxidase priming, which leads to an increase in fMLF-stimulated ROS production. This product is provided as a 10 mM solution in DMSO, suitable for various research applications.
  • Potent agonist for Toll-like Receptor 7 and 8
  • Induces pro-inflammatory cytokines in macrophages
  • Triggers NADPH oxidase priming
  • Provided as a 10 mM solution in DMSO
  • Suitable for research use
Catalog No. 50-003-42329