Learn More
Hach Company CL17 tubing - pump body to Y fitting and drain (order per foot) for Hach CL17 analyzer
Supplier:  Hach Company 4425300
Encompass
Replacement C-Flex tubing used to connect a CL17 analyzer pump body to the colorimeter Y-fitting and the sample drain. Sold by the foot for on-site replacement and custom routing.
  • C-Flex construction for chemical compatibility and flexibility.
  • 0.093 in outer diameter and 0.031 in inner diameter.
  • Sold per foot to allow custom-length installations.
  • Connects pump body, Y-fitting, and sample drain for CL17 analyzers.
  • Suitable for routine maintenance and tubing replacement.
Catalog No. NC3036731