Learn More
Medchemexpress LLC CL2-SN-38 | 1036969-20-6 | 99.2% | 1627.79 | C82H106N12O23 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY126350100MG
Encompass_Preferred
CL2-SN-38 is a cleavable linker-SN-38 conjugate intended for antibody-drug conjugate research. The molecule delivers the topoisomerase I inhibitor SN-38 linked by a cleavable spacer designed to release the active payload after antibody-mediated internalization. The product is supplied as a high-purity research reagent with documented solubility and storage recommendations.
  • Provides SN-38 payload for ADC development.
  • Features a cleavable linker for intracellular release.
  • Suitable for conjugation to antibodies targeting tumor antigens.
  • High purity for reproducible research results.
  • Soluble in DMSO at high concentration; in vivo formulations documented.
  • Supplied in small mg-scale quantities for laboratory use.
Catalog No. 50-002-03469