Learn More
Medchemexpress LLC Cl2a (PEG8- and triazole-containing PABC-peptide-mc linker) | 2616704-22-2 | 98.2% | 1062.21 | C50H79N9O16 | 50 MG

Supplier: Medchemexpress LLC HY12894550MG
CL2A is a cleavable PEG8- and triazole-containing PABC-peptide-mc linker used in antibody-drug conjugate research. It is pH-sensitive, enabling a bystander effect, and conjugates to antibodies through a disulfide bond at cysteine residues. Intended for research use only.
- Cleavable via pH sensitivity enabling bystander effect.
- Conjugates to antibody cysteine residues via a disulfide bond.
- Formula C50H79N9O16; molecular weight 1062.21.
- Purity typically 98.2% (reported batch value).
- Appearance colorless to light yellow solid-liquid mixture.
- Recommended storage: -20°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
- Available in small research sizes, including 50 MG.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.