Learn More
Medchemexpress LLC HY-128946 10mg , CL2A-SN-38 CAS:1279680-68-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12894610MG
Encompass_Preferred
HY-128946 10mg CL2A-SN-38 CAS: 1279680-68-0 CL2A-SN-38 is a drug-linker conjugate composed of a potent a DNA Topoisomerase I inhibitor SN-38 and a linker CL2A to make antibody drug conjugate (ADC). CL2A-SN-38 provides significant and specific antitumor effects against a range of human solid tumor types. CL2A is a nonclaevable complicated PEG8- and triazole-containing PABC-peptide-mc linker. CL2A is cleavable through pH sensitivity, giving rise to bystander effect, and binds the antibody at a cysteine residue via a disulfide bond. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9059